ContactUsLinkComponent
Thermometer similar to bs 593, b110c/total, 50+110:0,2â°c, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale 1 * 1 items
Catalog # AMARA403214-CB
Supplier:  Amarell
Thermometer similar to bs 593, b110c/total, 50+110:0,2â°c, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale 1 * 1 items
Catalog # AMARA403214-CB
Supplier:  Amarell
Frequently Bought Together