ContactUsLinkComponent
Thermometer similar to BS 593, A100C/total, 70+100:0,1C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA403206-CB
Supplier:  Amarell
Thermometer similar to BS 593, A100C/total, 70+100:0,1C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA403206-CB
Supplier:  Amarell
Frequently Bought Together