ContactUsLinkComponent
Saccharimeter Brix, 0-30:1/1Brix, with correction scale, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 20C, 300m m long 1 * 1 items
Catalog # AMARH846552-FL
Supplier:  Amarell
Saccharimeter Brix, 0-30:1/1Brix, with correction scale, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 20C, 300m m long 1 * 1 items
Catalog # AMARH846552-FL
Supplier:  Amarell
Frequently Bought Together