Thermometer similar to BS 1365, SB135C/total, 125+135:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402568-CB
Supplier: Amarell
Frequently Bought Together