ContactUsLinkComponent
Thermometer similar to BS 1365, SB65C/total, 55+65:0,1°C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402554-CB
Supplier:  Amarell
Thermometer similar to BS 1365, SB65C/total, 55+65:0,1°C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402554-CB
Supplier:  Amarell
Frequently Bought Together