ContactUsLinkComponent
Saccharimeter Brix, 10-20:0,1Brix, with correction scale, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 20C, 300 mm long 1 * 1 items
Catalog # AMARH845052-FL
Supplier:  Amarell
Saccharimeter Brix, 10-20:0,1Brix, with correction scale, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 20C, 300 mm long 1 * 1 items
Catalog # AMARH845052-FL
Supplier:  Amarell
Frequently Bought Together