ContactUsLinkComponent
Thermometer similar to BS 1365, SA105C/80, 45+105:0,5°C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 215mm long, immersion 80mm, durable pigment, null
  AMARA402522-CB
 :  Amarell
Thermometer similar to BS 1365, SA105C/80, 45+105:0,5°C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 215mm long, immersion 80mm, durable pigment, null
  AMARA402522-CB
 :  Amarell
Frequently Bought Together