ContactUsLinkComponent
Thermometer similar to BS 1365, SA105C/total, 45+105:0,5°C, solid stem, white backed, red special liquid, 215mm long, total immersio n, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA402502-FL
Supplier:  Amarell
Thermometer similar to BS 1365, SA105C/total, 45+105:0,5°C, solid stem, white backed, red special liquid, 215mm long, total immersio n, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA402502-FL
Supplier:  Amarell
Frequently Bought Together