ContactUsLinkComponent
Thermometer similar to BS 1365, SA105C/total, 45+105:0,5°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 215mm long, total immersion, durable pigment, null
Catalog # AMARA402502-CB
Supplier:  Amarell
Thermometer similar to BS 1365, SA105C/total, 45+105:0,5°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 215mm long, total immersion, durable pigment, null
Catalog # AMARA402502-CB
Supplier:  Amarell
Frequently Bought Together