Thermometer similar to BS 1704, D/75, 0+100:1°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liquid, 75mm immersion, 300mm long, durable pigment, null
Catalog # AMARA402106-CB
Supplier: Amarell
Frequently Bought Together