ContactUsLinkComponent
Thermo-hydrometer for milk and skim milk, similar to DIN 10290 (Gerber), with therm. +10+30:0,5C with red or blue special liquid, 1 ,020-1,040:0,0005g/cm , ref. temp. 20C, reading above, 320mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH841040-FL
Supplier:  Amarell
Thermo-hydrometer for milk and skim milk, similar to DIN 10290 (Gerber), with therm. +10+30:0,5C with red or blue special liquid, 1 ,020-1,040:0,0005g/cm , ref. temp. 20C, reading above, 320mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH841040-FL
Supplier:  Amarell
Frequently Bought Together