ContactUsLinkComponent
Thermometer similar to BS 1900, SR6/102C, 84+102:0,05°C, length 480mm, total immersion, solid stem, white backed, capillary tube sp ecially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401938-CB
Supplier:  Amarell
Thermometer similar to BS 1900, SR6/102C, 84+102:0,05°C, length 480mm, total immersion, solid stem, white backed, capillary tube sp ecially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401938-CB
Supplier:  Amarell
Frequently Bought Together