ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 26d, solid stem, white backed, -30-0:0,1°C, length 400mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400210-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 26d, solid stem, white backed, -30-0:0,1°C, length 400mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400210-CB
Supplier:  Amarell
Frequently Bought Together