ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 12d, solid stem, white backed, 130+160:0,1°C, length 420mm, imm. 100mm, capillary tube speciall y coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400080-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 12d, solid stem, white backed, 130+160:0,1°C, length 420mm, imm. 100mm, capillary tube speciall y coated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400080-CB
Supplier:  Amarell
Frequently Bought Together