ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 8d, solid stem, white backed, 70+130:0,2°C, length 380mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
  AMARA400050-CB
 :  Amarell
Thermometer similar to S.T.P.T.C. T 8d, solid stem, white backed, 70+130:0,2°C, length 380mm, imm. 100mm, capillary tube specially coated inside, with non-wetting blue special liquid, durable pigment, null
  AMARA400050-CB
 :  Amarell
Frequently Bought Together