ContactUsLinkComponent
TSB with neutralizer, CASO, Casein Soybean Digest Broth with 0.3% Lecithin, 3% Tween, 0.1% Histidin and 0.5% Thiosulfate, SCDB 1 * 20 items
Catalog # HEIP1462830020
Supplier:  MERCK LIFE SCIENCE
TSB with neutralizer, CASO, Casein Soybean Digest Broth with 0.3% Lecithin, 3% Tween, 0.1% Histidin and 0.5% Thiosulfate, SCDB 1 * 20 items
Catalog # HEIP1462830020
Supplier:  MERCK LIFE SCIENCE
Frequently Bought Together