Agilent InfinityLab Poroshell 120 EC-C18, 2.1 x 150 mm, 2.7 µm, 1000 bar 1 * 1 items
Catalog # HEWL693575-902
Supplier: Agilent
Agilent InfinityLab Poroshell 120 EC-C18, 2.1 x 150 mm, 2.7 µm, 1000 bar 1 * 1 items
Catalog # HEWL693575-902
Supplier: Agilent
Frequently Bought Together
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Avantor® ACE® C8, HPLC Columns, 5 µm
Supplier: Avantor
Screw neck vials, N 18 and N 20
Supplier: MACHEREY-NAGEL
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Supplier: Avantor
Glass crimp top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
TOC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
FlashPure Select Silica Cartridges
Supplier: BUCHI
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
UHPLC columns, InfinityLab Poroshell 120
Supplier: Agilent
Closures for screw neck headspace vials, Chromacol™
Supplier: CHROMACOL
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience
Microvolume syringes 10 µl
Supplier: Trajan Scientific and Medical