ContactUsLinkComponent
Jet Weaver mixer kit, 100 µL volume, for 1290 Infinity quaternary and 1290 Infinity II flexible pump, includes mixer assembly and co nnecting capillaries 1 * 1 items
Catalog # HEWLG4204-68100
Supplier:  Agilent
Jet Weaver mixer kit, 100 µL volume, for 1290 Infinity quaternary and 1290 Infinity II flexible pump, includes mixer assembly and co nnecting capillaries 1 * 1 items
Catalog # HEWLG4204-68100
Supplier:  Agilent
Frequently Bought Together