ContactUsLinkComponent
SILAC Protein Quantitation Kit - DMEM is used for the specific analysis of protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC). 1 * 1 KIT
Catalog # PIERA33969
SILAC Protein Quantitation Kit - DMEM is used for the specific analysis of protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC). 1 * 1 KIT
Catalog # PIERA33969
Frequently Bought Together