ContactUsLinkComponent
READYPLATE 55 KIT PLATE COUNT AGAR PLATE COUNT AGAR IN 55MM PLATE WITH EZ PAK MEMBRANE FILTER 1 * 1 items
Catalog # HEIP1467640150
Supplier:  MERCK LIFE SCIENCE
READYPLATE 55 KIT PLATE COUNT AGAR PLATE COUNT AGAR IN 55MM PLATE WITH EZ PAK MEMBRANE FILTER 1 * 1 items
Catalog # HEIP1467640150
Supplier:  MERCK LIFE SCIENCE
Frequently Bought Together