ContactUsLinkComponent
Graduated 5 mL Transfer Pipettes, large bulb. Length 21,7 cm. Stem Diameter 3,0 mm. Overall Capacity 13,7 mL. Bulb Draw 7,4 mL. 19 drops per mL. 1 * 2.500 items
Catalog # SAMC728NL
Supplier:  SAMCO
undefined
Graduated 5 mL Transfer Pipettes, large bulb. Length 21,7 cm. Stem Diameter 3,0 mm. Overall Capacity 13,7 mL. Bulb Draw 7,4 mL. 19 drops per mL. 1 * 2.500 items
Catalog # SAMC728NL
Supplier:  SAMCO
Frequently Bought Together