ContactUsLinkComponent
General Purpose Transfer Pipettes, 4 B/B Pet (Blood Bank). Length 10,3 cm. Stem Diameter 2,5 mm. Overall Capacity 3,1 mL. Bulb Draw 2,2 mL. 22 drops per mL. 1 * 4.000 items
Catalog # SAMC334-11
Supplier:  SAMCO
General Purpose Transfer Pipettes, 4 B/B Pet (Blood Bank). Length 10,3 cm. Stem Diameter 2,5 mm. Overall Capacity 3,1 mL. Bulb Draw 2,2 mL. 22 drops per mL. 1 * 4.000 items
Catalog # SAMC334-11
Supplier:  SAMCO
Frequently Bought Together