ContactUsLinkComponent
General Purpose Transfer Pipettes, Small Bulb. Length 12,7 cm. Stem Diameter 2,5 mm. Overall Capacity 13,7 mL. Bulb Draw 7,4 mL. 24 drops per mL. 1 * 5.000 items
Catalog # SAMC325-11
Supplier:  SAMCO
undefined
General Purpose Transfer Pipettes, Small Bulb. Length 12,7 cm. Stem Diameter 2,5 mm. Overall Capacity 13,7 mL. Bulb Draw 7,4 mL. 24 drops per mL. 1 * 5.000 items
Catalog # SAMC325-11
Supplier:  SAMCO
Frequently Bought Together