Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Bromomethyl methyl ether

Bromomethyl methyl ether

Bottle, 5 g
Catalog # APOSOR11138-5G
Flammable
Irritant
Health
Supplier:   Apollo Scientific
Bromomethyl methyl ether
Catalog # APOSOR11138-5G
Supplier:  Apollo Scientific
CAS Number:  13057-17-5
Bromomethyl methyl ether

Bottle, 5 g

CAS Number:  13057-17-5
Price
697,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:13057-17-5
  • Formula:C₂H₅BrO
  • MDL Number:MFCD00000171
  • Molecular Weight:124.97 g/mol
  • Boiling Pt:27 °C
  • Melting Pt:87 °C
  • Density:1.531 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
  • UN:1992
  • ADR:3+6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.