6-Fluoroquinoline
Bottle, 5 g
Katalogové číslo APOSPC6908-5G
Dodavatel: Apollo Scientific
Přejít na:
Product Details & Documents
- Číslo CAS:396-30-5
- Vzorec:C₉H₆FN
- MDL číslo:MFCD01685512
- Molecular Weight:147.15 g/mol
- Bod varu:124 °C/30mmHg
- Hustota:1.209 g/cm³ (25 °C)
- Teplota pro skladování:Refrigerator
Specifikace
Specifikace
- Pack type:Bottle
- Pk:5 g
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



