Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
6-Fluoroquinoline

6-Fluoroquinoline

Bottle, 1 g
Catalog # APOSPC6908-1G
Corrosive
Irritant
Supplier:   Apollo Scientific
6-Fluoroquinoline
Catalog # APOSPC6908-1G
Supplier:  Apollo Scientific
CAS Number:  396-30-5
6-Fluoroquinoline

Bottle, 1 g

CAS Number:  396-30-5
Price
615,00 Kč
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:396-30-5
  • Formula:C₉H₆FN
  • MDL Number:MFCD01685512
  • Molecular Weight:147.15 g/mol
  • Boiling Pt:124 °C/30mmHg
  • Density:1.209 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications