Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Ethyl-4,4-difluorocyclohexanecarboxylate

Bottle, 5 g
Catalog # APOSPC0689-5G
Irritant
Supplier:   Apollo Scientific
Ethyl-4,4-difluorocyclohexanecarboxylate
Catalog # APOSPC0689-5G
Supplier:  Apollo Scientific
CAS Number:  178312-47-5
Ethyl-4,4-difluorocyclohexanecarboxylate

Bottle, 5 g

CAS Number:  178312-47-5
Price
902,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Ethyl 4,4-difluorocyclohexanecarboxylate
  • CAS Number:178312-47-5
  • Formula:C₉H₁₄F₂O₂
  • MDL Number:MFCD03788494
  • Molecular Weight:192.21 g/mol
  • Boiling Pt:84…86 °C/8mmHg
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications