Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
2-(4-Fluorophenoxy)-3-nitropyridine 97%

2-(4-Fluorophenoxy)-3-nitropyridine 97%

Bottle, 5 g
Catalog # APOSPC4107F-5G
Supplier:   Apollo Scientific
2-(4-Fluorophenoxy)-3-nitropyridine 97%
Catalog # APOSPC4107F-5G
Supplier:  Apollo Scientific
CAS Number:  147143-58-6
2-(4-Fluorophenoxy)-3-nitropyridine 97%

Bottle, 5 g

CAS Number:  147143-58-6
Price
7 010,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:147143-58-6
  • Formula:C₁₁H₇FN₂O₃
  • MDL Number:MFCD00067801
  • Molecular Weight:234.19 g/mol
  • Melting Pt:98…99 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.