Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
8-Fluoroquinoline

8-Fluoroquinoline

Bottle, 1 g
Catalog # APOSPC3205-1G
Irritant
Supplier:   Apollo Scientific
8-Fluoroquinoline
Catalog # APOSPC3205-1G
Supplier:  Apollo Scientific
CAS Number:  394-68-3
8-Fluoroquinoline

Bottle, 1 g

CAS Number:  394-68-3
Price
615,00 Kč
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:394-68-3
  • Formula:C₉H₆FN
  • MDL Number:MFCD01685514
  • Molecular Weight:147.1500 g/mol
  • Boiling Pt:148…150 °C/30mmHg
  • Density:1.215 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications