Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
N,O-Di-boc-hydroxylamine

N,O-Di-boc-hydroxylamine

Bottle, 1 g
Catalog # APOSOR46242-1G
Irritant
Supplier:   Apollo Scientific
N,O-Di-boc-hydroxylamine
Catalog # APOSOR46242-1G
Supplier:  Apollo Scientific
CAS Number:  85006-25-3
N,O-Di-boc-hydroxylamine

Bottle, 1 g

CAS Number:  85006-25-3
Price
615,00 Kč
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  N,O-Di-Boc-hydroxylamine, tert-Butyl N-(tert-Butoxycarbonyloxy)carbamate, N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl (tert-butoxycarbonyl)oxycarbamate, N-(tert-Butoxycarbonyloxy)carbamic acid tert-Butyl ester
  • CAS Number:85006-25-3
  • Formula:C₁₀H₁₉NO₅
  • MDL Number:MFCD00034797
  • Molecular Weight:233.264 g/mol
  • Melting Pt:67…70 °C
  • Storage Temperature:Freezer
  • EINECS:285-055-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications