HT-2 Toxin standard solution
HT-2 Toxin, 100 µg/ml in acetonitrile, Bottle, 1 mL
Katalogové číslo APOSBIMS115-1ML
Dodavatel: Apollo Scientific
HT-2 Toxin standard solution
Katalogové číslo APOSBIMS115-1ML
Dodavatel: Apollo Scientific
Číslo CAS:
Jump to:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:1 mL
- Description:HT-2 Toxin
- Cat. No.:APOSBIMS115-1ML
- Formula:C₂₂H₃₂O₈
- Description:HT-2 Toxin
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:26934-87-2
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



