Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Imidazole

Imidazole

Bottle, 500 g
Catalog # APOSBID0565-500G
Corrosive
Irritant
Health
Supplier:   Apollo Scientific
Imidazole
Catalog # APOSBID0565-500G
Supplier:  Apollo Scientific
CAS Number:  288-32-4
Imidazole

Bottle, 500 g

CAS Number:  288-32-4
Price
656,00 Kč
Log in for availability
1 * 500 g
Product Details & Documents

Biological buffer - useful range 6.2-7.8

  • Assay (by titration) 99%
  • White crystals
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Boiling Pt:235 °C
  • Melting Pt:85…91 °C
  • Density:1.03 g/cm³ (25 °C)
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,II

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications