Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
(S)-(+)-Camptothecin

(S)-(+)-Camptothecin

Bottle, 5 g
Catalog # APOSBIC8442-5G
Toxic
Supplier:   Apollo Scientific
(S)-(+)-Camptothecin
Catalog # APOSBIC8442-5G
Supplier:  Apollo Scientific
CAS Number:  7689-03-4
(S)-(+)-Camptothecin

Bottle, 5 g

CAS Number:  7689-03-4
Price
3 080,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents

A potent antitumor antibiotic.

Synonyms:  (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
  • CAS Number:7689-03-4
  • Formula:C₂₀H₁₆N₂O₄
  • MDL Number:MFCD00081076
  • Molecular Weight:348.36 g/mol
  • Melting Pt:260 °C (dec.)
  • Storage Temperature:Refrigerator
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.