Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
(S)-(+)-Camptothecin

(S)-(+)-Camptothecin

Bottle, 10 g
Catalog # APOSBIC8442-10G
Toxic
Supplier:   Apollo Scientific
(S)-(+)-Camptothecin
Catalog # APOSBIC8442-10G
Supplier:  Apollo Scientific
CAS Number:  7689-03-4
(S)-(+)-Camptothecin

Bottle, 10 g

CAS Number:  7689-03-4
Price
4 920,00 Kč
Log in for availability
1 * 10 g
Product Details & Documents

A potent antitumor antibiotic.

Synonyms:  (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
  • CAS Number:7689-03-4
  • Formula:C₂₀H₁₆N₂O₄
  • MDL Number:MFCD00081076
  • Molecular Weight:348.36 g/mol
  • Melting Pt:260 °C (dec.)
  • Storage Temperature:Refrigerator
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications