ContactUsLinkComponent
Volts: 400V - Phase: 3phase Ampere: 3 A Watt: 1750 W Airflow: 238 m /h Vacuum pressure: 2819 mmH20 Recovery Capacity: -28L - For dry recovery only. -15L - With paper filter bag. Dimensions: H109cmxW50cmxL34cm Weight: 52 Kg Noise Level: 64.00 dB(A) 1 * 1 items
Catalog # TIGV110985B3
Supplier:  TIGER-VAC
Volts: 400V - Phase: 3phase Ampere: 3 A Watt: 1750 W Airflow: 238 m /h Vacuum pressure: 2819 mmH20 Recovery Capacity: -28L - For dry recovery only. -15L - With paper filter bag. Dimensions: H109cmxW50cmxL34cm Weight: 52 Kg Noise Level: 64.00 dB(A) 1 * 1 items
Catalog # TIGV110985B3
Supplier:  TIGER-VAC
Frequently Bought Together