Order Entry
Czech Republic
ContactUsLinkComponent
25PPMH2S-100PPMCO-2.2%CH4-18%O2INN2 58L 1 * 1 items
Catalog # BWTE4GA18X22W25H58
Supplier:  BW TECHNOLOGIES
25PPMH2S-100PPMCO-2.2%CH4-18%O2INN2 58L 1 * 1 items
Catalog # BWTE4GA18X22W25H58
Supplier:  BW TECHNOLOGIES
Frequently Bought Together