ContactUsLinkComponent
Fixed high intensity multiplexing LEDs with peak band passes of Red (635 nm), Green (520 nm), Blue (465 nm) 1 * 1 items
Catalog # SYNOGM-HILED-RGB
Fixed high intensity multiplexing LEDs with peak band passes of Red (635 nm), Green (520 nm), Blue (465 nm) 1 * 1 items
Catalog # SYNOGM-HILED-RGB
Frequently Bought Together