Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
 

Acetazolamide, Reference Standards

Acetazolamide Related Compound E, USP reference standard, 25 mg
  1005059.
 :   USP
Acetazolamide, Reference Standards
  1005059.
 :  USP
 :  
Acetazolamide, Reference Standards

Acetazolamide Related Compound E, USP reference standard, 25 mg

 :  
 
30 680,00 Kč
   
1 * 25 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

  • Bal.:
    25 mg
  • Standard:
    USP reference standard
  • Popis:
    Acetazolamide Related Compound E
  • Popis:
    Acetazolamide Related Compound E
  • Cat. No.:
    1005059.
  • Synonym(s):
    5-Acetamido-1,3,4-thiadiazole-2-sulfonic acid potassium salt

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.