Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Acetazolamide, Reference Standards

Acetazolamide, Reference Standards

Acetazolamide Related Compound D, USP reference standard, 25 mg
Catalog # 1005048.
Supplier:   USP
Acetazolamide, Reference Standards
Catalog # 1005048.
Supplier:  USP
CAS Number:  
Acetazolamide, Reference Standards

Acetazolamide Related Compound D, USP reference standard, 25 mg

CAS Number:  
Price
28 370,00 Kč
Log in for availability
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pk:
    25 mg
  • Standards:
    USP reference standard
  • Description:
    Acetazolamide Related Compound D
  • Cat. No.:
    1005048.
  • Description:
    Acetazolamide Related Compound D
  • Synonym(s):
    5-Amino-1,3,4-thiadiazole-2-sulfonamide
  • CAS No.:
    14949-00-9

Specifications