ContactUsLinkComponent
1.5 Litre Commercial Blender Complete with BPA-free Copolyester Stackable Container, Toggle Switch Controls, 2 HP, 1500W 2-speed motor, 24,000 max RPM (CE Approved ) 1 * 1 items
Catalog # WARITBB145E
Supplier:  WARING BLENDOR
1.5 Litre Commercial Blender Complete with BPA-free Copolyester Stackable Container, Toggle Switch Controls, 2 HP, 1500W 2-speed motor, 24,000 max RPM (CE Approved ) 1 * 1 items
Catalog # WARITBB145E
Supplier:  WARING BLENDOR
Frequently Bought Together