ContactUsLinkComponent
VarioPrep HPLC guard column cartridge (short) VP 10/16 NUCLEOSIl C18 Length: 10 mm, ID: 16 mm pack of 2 - compatible with VarioPrep HPLC column with ID: 16 mm or ID: 21 mm - requires VP guard column holder 16 mm (short version, REF: 718256) 1 * 2 items
Catalog # MANA715360.160
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC guard column cartridge (short) VP 10/16 NUCLEOSIl C18 Length: 10 mm, ID: 16 mm pack of 2 - compatible with VarioPrep HPLC column with ID: 16 mm or ID: 21 mm - requires VP guard column holder 16 mm (short version, REF: 718256) 1 * 2 items
Catalog # MANA715360.160
Supplier:  MACHEREY-NAGEL
Frequently Bought Together