ContactUsLinkComponent
Recirculating Chiller F-325 2500W at 15°C, Display, 230V. The F-325 is an easy to use recirculating chiller designed for the Rotavapor R-220 Pro. It features temperature lock, ECO-mode and temperature control in conjunction with the Rotavapor R-220 P 1 * 1 items
Catalog # BUCH11F32501
Supplier:  BUCHI
Recirculating Chiller F-325 2500W at 15°C, Display, 230V. The F-325 is an easy to use recirculating chiller designed for the Rotavapor R-220 Pro. It features temperature lock, ECO-mode and temperature control in conjunction with the Rotavapor R-220 P 1 * 1 items
Catalog # BUCH11F32501
Supplier:  BUCHI
Frequently Bought Together