ContactUsLinkComponent
EC HPLC column EC 150/4 NUCLEOSIL 100-3 C18 Length: 150 mm, ID: 4 mm pack of 1 1 * 1 items
Katalogové číslo MANA720949.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 150/4 NUCLEOSIL 100-3 C18 Length: 150 mm, ID: 4 mm pack of 1 1 * 1 items
Katalogové číslo MANA720949.40
Supplier:  MACHEREY-NAGEL
Frequently Bought Together