ContactUsLinkComponent
Fluorocult DEV Lactose Peptone Broth is particulary suited for the enrichment and determinato of the titre of coliform bacteria in the bacterlogical analysis of water. E. coli shows a positive fluorescence under UV light (366 nm) A positive indole r 1 * 2 kg
Catalog # FCHKFCM-247
Supplier:  FOODCHEK SYSTEMS
Fluorocult DEV Lactose Peptone Broth is particulary suited for the enrichment and determinato of the titre of coliform bacteria in the bacterlogical analysis of water. E. coli shows a positive fluorescence under UV light (366 nm) A positive indole r 1 * 2 kg
Catalog # FCHKFCM-247
Supplier:  FOODCHEK SYSTEMS
Frequently Bought Together