ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H L) with our bright and photostable CF™ 770. To minimize cross-reactivity, the antibody has been adsorbed against  bovine, chicken, goat, guinea pig, Syrian hamster, 1 * 50 µl
Catalog # 20484-50UL
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H L) with our bright and photostable CF™ 770. To minimize cross-reactivity, the antibody has been adsorbed against  bovine, chicken, goat, guinea pig, Syrian hamster, 1 * 50 µl
Catalog # 20484-50UL
Supplier:  Biotium
Frequently Bought Together