ContactUsLinkComponent
Liquid electrolyte comb. electrode with integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum junction, elec trolyte L 200, sphere membrane, A-glass, coax screw plug head, length 120 mm, 12 mm Ø, -30...+100°C, 0...14 pH 1 * 1 items
Catalog # SCOI285110260
Supplier:  SI Analytics
Liquid electrolyte comb. electrode with integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum junction, elec trolyte L 200, sphere membrane, A-glass, coax screw plug head, length 120 mm, 12 mm Ø, -30...+100°C, 0...14 pH 1 * 1 items
Catalog # SCOI285110260
Supplier:  SI Analytics