ContactUsLinkComponent
Low maintenance redox-combination electrode with NTC 30K, glass shaft, KPG annular gap junction, Referid®-electrolyte, Silamid®-ref erence system, sensor platinum disk 6 mm Ø, Memosens® screw plug head, length 225 mm, 12 mm Ø, -5...+100 °C 1 * 1 items
Catalog # SCOI285111110
Supplier:  SI Analytics
Low maintenance redox-combination electrode with NTC 30K, glass shaft, KPG annular gap junction, Referid®-electrolyte, Silamid®-ref erence system, sensor platinum disk 6 mm Ø, Memosens® screw plug head, length 225 mm, 12 mm Ø, -5...+100 °C 1 * 1 items
Catalog # SCOI285111110
Supplier:  SI Analytics