ContactUsLinkComponent
Liquid electrolyte comb. electrode with integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum junction, elec trolyte L 300, sphere membrane, H-glass, coax screw plug head, length 120 mm, 12 mm Ø, 0...+135°C, 0...14 pH 1 * 1 items
Catalog # SCOI285110370
Supplier:  SI Analytics
Liquid electrolyte comb. electrode with integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum junction, elec trolyte L 300, sphere membrane, H-glass, coax screw plug head, length 120 mm, 12 mm Ø, 0...+135°C, 0...14 pH 1 * 1 items
Catalog # SCOI285110370
Supplier:  SI Analytics