ContactUsLinkComponent
pH combination electrode, screw plug head (ATEX II 1/2G), low maintenance, Pt 1000, glass shaft, KPG annular gap diaphragm, Referid® -electrolyte, Silamid®-reference, dome membrane, H-glass, VP plug head, length 120 mm, 12 mm Ø, 0...+100 °C, 2...13 p 1 * 1 items
Catalog # SCOI285094780
Supplier:  SI Analytics
pH combination electrode, screw plug head (ATEX II 1/2G), low maintenance, Pt 1000, glass shaft, KPG annular gap diaphragm, Referid® -electrolyte, Silamid®-reference, dome membrane, H-glass, VP plug head, length 120 mm, 12 mm Ø, 0...+100 °C, 2...13 p 1 * 1 items
Catalog # SCOI285094780
Supplier:  SI Analytics