ContactUsLinkComponent
Liquid electrolyte comb. electrode with Pt 1000 and integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum ju nction, electrolyte L 300, sphere membrane, H-glass, VP screw plug head, length 120 mm, 12 mm Ø, 0...+135 °C, 0...14 p 1 * 1 items
Catalog # SCOI285110410
Supplier:  SI Analytics
Liquid electrolyte comb. electrode with Pt 1000 and integrated plastic adapter for hose connection incl. Pg13.5 thread, platinum ju nction, electrolyte L 300, sphere membrane, H-glass, VP screw plug head, length 120 mm, 12 mm Ø, 0...+135 °C, 0...14 p 1 * 1 items
Catalog # SCOI285110410
Supplier:  SI Analytics